Assay Design
Gene Transcripts
- Gene Symbol
- Duoxa1
- Entrez Gene ID
-
213696
- Gene Name
- dual oxidase maturation factor 1
- Gene Aliases
-
BC019755, Nip, Nip1
- Chromosome Location
-
Chr.2: 122303540 - 122313744 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC019755.1 |
- |
4 - 5 |
241 |
139 |
View Full Product Details