Assay Design



Gene Transcripts

Gene Symbol
Kri1
Entrez Gene ID
215194
Gene Name
KRI1 homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.9: 21273457 - 21287969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145416.3 NP_663391.2 3 - 4 292 77
GenBank mRNA AK051997.1 - 3 - 4 293 77
BC021438.1 - 3 - 4 285 77
CJ069606.1 - 3 - 4 294 77

View Full Product Details