Assay Design



Gene Transcripts

Gene Symbol
Nle1
Entrez Gene ID
217011
Gene Name
notchless homolog 1 (Drosophila)
Gene Aliases
AL022765, BC018399, Nle, l11Jus1, l11Jus4
Chromosome Location
Chr.11: 82900768 - 82908445 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145431.2 NP_663406.2 2 - 3 208 101
GenBank mRNA AK141230.1 - 2 - 3 183 101
AK170853.1 - 2 - 3 181 101
BC018399.1 - 2 - 3 199 101
BC035421.1 - 2 - 3 199 101
BY149184.1 - 2 - 3 209 101

View Full Product Details