Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145434.4 | NP_663409.2 | 4 - 5 | 1236 | 72 |
GenBank mRNA | AK041047.1 | - | 4 - 5 | 1092 | 72 |
AK137398.1 | - | 4 - 5 | 1123 | 72 | |
AK137582.1 | - | 4 - 5 | 1101 | 72 | |
AK146430.1 | - | 4 - 5 | 1193 | 72 | |
AK154931.1 | - | 4 - 5 | 1237 | 72 | |
AK155597.1 | - | 4 - 5 | 1237 | 72 | |
BC008989.1 | - | 4 - 5 | 681 | 72 |