Assay Design



Gene Transcripts

Gene Symbol
Eif2b2
Entrez Gene ID
217715
Gene Name
eukaryotic translation initiation factor 2B, subunit 2 beta
Gene Aliases
AA409345, C85417, EIF-2Bbeta, EIF2B
Chromosome Location
Chr.12: 85219475 - 85226628 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145445.3 NP_663420.1 4 - 5 647 72
GenBank mRNA AK049731.1 - 4 - 5 648 72
AK076195.1 - 4 - 5 643 72
BC003326.1 - 4 - 5 679 72

View Full Product Details