Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145445.3 | NP_663420.1 | 6 - 7 | 877 | 97 |
GenBank mRNA | AK049731.1 | - | 6 - 7 | 878 | 97 |
AK076195.1 | - | 6 - 7 | 873 | 97 | |
AK195610.1 | - | 2 - 3 | 169 | 97 | |
AK206706.1 | - | 2 - 3 | 169 | 97 | |
AK213656.1 | - | 2 - 3 | 169 | 97 | |
BC003326.1 | - | 6 - 7 | 909 | 97 |