Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166453.1 | NP_001159925.1 | 2 - 3 | 205 | 126 |
NM_001166454.2 | NP_001159926.1 | 2 - 3 | 200 | 126 | |
NM_145629.2 | NP_663604.1 | 2 - 3 | 319 | 126 | |
GenBank mRNA | AB182243.1 | - | 1 - 2 | 74 | 126 |
AK146087.1 | - | 2 - 3 | 130 | 126 | |
AK146455.1 | - | 2 - 3 | 122 | 126 | |
AK160501.1 | - | 2 - 3 | 148 | 126 | |
AK165540.1 | - | 2 - 3 | 205 | 126 | |
BB872046.1 | - | 2 - 3 | 205 | 126 | |
BC005459.1 | - | 2 - 3 | 280 | 126 | |
BY020402.1 | - | 2 - 3 | 242 | 126 |