Assay Design



Gene Transcripts

Gene Symbol
Olah
Entrez Gene ID
99035
Gene Name
oleoyl-ACP hydrolase
Gene Aliases
AI607300, E230009B14Rik, Thedc1
Chromosome Location
Chr.2: 3341982 - 3397238 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145921.1 NP_666033.1 3 - 4 446 79
GenBank mRNA AK087571.1 - 3 - 4 466 79
AK136031.1 - 4 - 5 702 79
BC025001.1 - 3 - 4 446 79

View Full Product Details