Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010687.1 | NP_034817.1 | 11 - 12 | 1619 | 67 |
GenBank mRNA | AJ006278.1 | - | 11 - 12 | 1619 | 67 |
AK122328.1 | - | 12 - 13 | 2696 | 67 | |
AK133940.1 | - | 11 - 12 | 2104 | 67 | |
BC061506.1 | - | 10 - 11 | 1376 | 67 | |
BC079596.1 | - | 11 - 12 | 1902 | 67 | |
BC100399.1 | - | 11 - 12 | 1682 | 67 | |
BC125487.1 | - | 11 - 12 | 1585 | 67 | |
BC125489.1 | - | 11 - 12 | 1585 | 67 | |
HM141820.1 | - | 9 - 10 | 1254 | 67 |