Assay Design



Gene Transcripts

Gene Symbol
Mbd1
Entrez Gene ID
17190
Gene Name
methyl-CpG binding domain protein 1
Gene Aliases
Cxxc3, PCM1
Chromosome Location
Chr.18: 74267615 - 74282734 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013594.2 NP_038622.2 3 - 4 392 108
GenBank mRNA AF072240.1 - 3 - 4 395 108
AK004624.2 - 3 - 4 393 108
AK156401.1 - 3 - 4 407 108
AK162858.1 - 3 - 4 392 108
BC069837.1 - 3 - 4 408 108

View Full Product Details