Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190984.1 | NP_001177913.1 | 4 - 5 | 491 | 80 |
NM_001190985.1 | NP_001177914.1 | 3 - 4 | 709 | 80 | |
NM_021295.3 | NP_067270.1 | 4 - 5 | 525 | 80 | |
GenBank mRNA | AJ294535.1 | - | 4 - 5 | 518 | 80 |
BC038649.1 | - | 1 - 2 | 30 | 80 | |
BC058560.1 | - | 4 - 5 | 491 | 80 | |
Y11550.1 | - | 4 - 5 | 490 | 80 | |
Y16518.1 | - | 3 - 4 | 669 | 80 |