Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021921.3 | NP_068740.3 | 3 - 4 | 760 | 66 |
GenBank mRNA | AF220195.1 | - | 3 - 4 | 479 | 66 |
AF310135.1 | - | 3 - 4 | 541 | 66 | |
AK014339.1 | - | 3 - 4 | 464 | 66 | |
AK158968.1 | - | 3 - 4 | 614 | 66 | |
BC029704.1 | - | 3 - 4 | 539 | 66 |