Assay Design



Gene Transcripts

Gene Symbol
Lbh
Entrez Gene ID
77889
Gene Name
limb-bud and heart
Gene Aliases
1810009F10Rik, 6720416L16Rik
Chromosome Location
Chr.17: 72918305 - 72941946 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029999.4 NP_084275.3 1 - 2 275 85
GenBank mRNA AF317517.1 - 1 - 2 258 85
AK020110.1 - 1 - 2 275 85
BC026827.1 - 1 - 2 257 85
BC052470.1 - 1 - 2 255 85

View Full Product Details