Assay Design



Gene Transcripts

Gene Symbol
Nrbp2
Entrez Gene ID
223649
Gene Name
nuclear receptor binding protein 2
Gene Aliases
BC011468
Chromosome Location
Chr.15: 76085589 - 76091594 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144847.1 NP_659096.1 11 - 12 947 69
GenBank mRNA BC011468.1 - 11 - 12 880 69
BC012437.1 - 11 - 12 947 69
BC025042.1 - 6 - 7 409 69
BC068117.1 - 16 - 17 1595 69

View Full Product Details