Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144851.5 | NP_659100.1 | 6 - 7 | 884 | 80 |
GenBank mRNA | AK028565.1 | - | 6 - 7 | 626 | 80 |
AK035581.1 | - | 6 - 7 | 720 | 80 | |
AK054521.1 | - | 7 - 8 | 1016 | 80 | |
AK088597.1 | - | 6 - 7 | 628 | 80 | |
AK138013.1 | - | 6 - 7 | 685 | 80 | |
BC023129.1 | - | 5 - 6 | 598 | 80 |