Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144860.2 | NP_659109.2 | 13 - 14 | 2646 | 68 |
GenBank mRNA | AK053035.1 | - | 6 - 7 | 869 | 68 |
AK080847.1 | - | 7 - 8 | 922 | 68 | |
AY147848.1 | - | 13 - 14 | 2145 | 68 | |
AY149907.1 | - | 13 - 14 | 1966 | 68 | |
AY245539.1 | - | 13 - 14 | 2646 | 68 | |
AY974091.1 | - | 13 - 14 | 1939 | 68 | |
BC011287.1 | - | 2 - 3 | 243 | 68 | |
BC083072.1 | - | 3 - 4 | 324 | 68 | |
BC131662.1 | - | 14 - 15 | 2112 | 68 |