Assay Design



Gene Transcripts

Gene Symbol
Klhl20
Entrez Gene ID
226541
Gene Name
kelch-like 20
Gene Aliases
AI504637, D930050H05Rik, Kleip, mKIAA4210
Chromosome Location
Chr.1: 161088375 - 161131508 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039482.1 NP_001034571.1 11 - 12 1899 90
GenBank mRNA AK032131.1 - 9 - 10 1387 90
AK046442.1 - 2 - 3 199 90
AK053102.1 - 11 - 12 1849 90
AK220547.1 - 11 - 12 1878 90
BC019571.1 - 11 - 12 1871 90
BC029801.1 - 5 - 6 622 90
BC060183.1 - 10 - 11 2573 90

View Full Product Details