Assay Design



Gene Transcripts

Gene Symbol
Gdap1l1
Entrez Gene ID
228858
Gene Name
ganglioside-induced differentiation-associated protein 1-like 1
Gene Aliases
Gdap1l
Chromosome Location
Chr.2: 163438419 - 163455324 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144891.2 NP_659140.2 2 - 3 453 53
GenBank mRNA AK140488.1 - 2 - 3 444 53
AK141371.1 - 2 - 3 445 53
AK164991.1 - 2 - 3 504 53
BB649122.1 - 2 - 3 469 53
BC019941.1 - 2 - 3 467 53

View Full Product Details