Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144900.2 | NP_659149.1 | 19 - 20 | 3010 | 68 |
GenBank mRNA | AK088000.1 | - | 19 - 20 | 2971 | 68 |
AK137001.1 | - | 19 - 20 | 2970 | 68 | |
AK148989.1 | - | 6 - 7 | 868 | 68 | |
AK159279.1 | - | 19 - 20 | 2967 | 68 | |
AK159306.1 | - | 19 - 20 | 2965 | 68 | |
AK177206.1 | - | - | 41 | 68 | |
AK181491.1 | - | 1 - 2 | 41 | 68 | |
AK191830.1 | - | 1 - 2 | 41 | 68 | |
BC010319.1 | - | 17 - 18 | 2595 | 68 | |
BC021496.1 | - | 19 - 20 | 2960 | 68 | |
BC023175.1 | - | 3 - 4 | 304 | 68 | |
BC023794.1 | - | 20 - 21 | 2905 | 68 | |
BC023867.1 | - | 2 - 3 | 306 | 68 | |
BC023964.1 | - | 19 - 20 | 2951 | 68 | |
BC025037.1 | - | 12 - 13 | 1810 | 68 | |
BC025618.1 | - | 19 - 20 | 2969 | 68 | |
BC025627.1 | - | 19 - 20 | 2960 | 68 | |
BC025811.1 | - | 19 - 20 | 2947 | 68 | |
BC030326.1 | - | 8 - 9 | 1307 | 68 | |
BC031176.1 | - | 6 - 7 | 868 | 68 | |
BC031389.1 | - | 3 - 4 | 429 | 68 | |
BC032187.1 | - | 19 - 20 | 3010 | 68 | |
BC033435.1 | - | 19 - 20 | 2906 | 68 | |
BC033471.1 | - | 19 - 20 | 2950 | 68 | |
BC034177.1 | - | 18 - 19 | 2715 | 68 | |
BC035352.1 | - | 18 - 19 | 2680 | 68 | |
BC042435.1 | - | 19 - 20 | 2960 | 68 |