Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290525.1 | NP_001277454.1 | 4 - 5 | 517 | 62 |
NM_144923.3 | NP_659172.1 | 4 - 5 | 603 | 62 | |
GenBank mRNA | AK153153.1 | - | 4 - 5 | 524 | 62 |
BC006617.1 | - | 4 - 5 | 543 | 62 | |
BC027279.1 | - | 4 - 5 | 603 | 62 |