Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252566.1 | NP_001239495.1 | 16 - 17 | 3204 | 67 |
NM_001252567.1 | NP_001239496.1 | 17 - 18 | 3063 | 67 | |
NM_144926.5 | NP_659175.1 | 17 - 18 | 3243 | 67 | |
GenBank mRNA | AK034088.1 | - | 17 - 18 | 3215 | 67 |
AK165738.1 | - | 16 - 17 | 3176 | 67 | |
BC011475.1 | - | 17 - 18 | 3049 | 67 | |
BC079573.1 | - | 17 - 18 | 2632 | 67 | |
BC096615.1 | - | 16 - 17 | 3116 | 67 |