Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285997.1 | NP_001272926.1 | 6 - 7 | 954 | 126 |
NM_001285998.1 | NP_001272927.1 | 6 - 7 | 954 | 126 | |
NM_145150.3 | NP_660132.2 | 6 - 7 | 954 | 126 | |
GenBank mRNA | AK029140.1 | - | 6 - 7 | 941 | 126 |
AK088576.1 | - | 6 - 7 | 926 | 126 | |
AK135778.1 | - | 4 - 5 | 579 | 126 | |
BC005475.1 | - | 6 - 7 | 888 | 126 | |
BC059808.1 | - | 6 - 7 | 899 | 126 |