Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285997.1 | NP_001272926.1 | 12 - 13 | 1690 | 84 |
NM_001285998.1 | NP_001272927.1 | 12 - 13 | 1699 | 84 | |
NM_145150.3 | NP_660132.2 | 12 - 13 | 1699 | 84 | |
GenBank mRNA | AF408435.1 | - | 4 - 5 | 458 | 84 |
AK088576.1 | - | 12 - 13 | 1671 | 84 | |
AK135778.1 | - | 10 - 11 | 1325 | 84 | |
AK190153.1 | - | 1 - 2 | 36 | 84 | |
BC005475.1 | - | 12 - 13 | 1624 | 84 | |
BC059808.1 | - | 12 - 13 | 1644 | 84 |