Assay Design



Gene Transcripts

Gene Symbol
Gbp7
Entrez Gene ID
229900
Gene Name
guanylate binding protein 7
Gene Aliases
9830147J24Rik
Chromosome Location
Chr.3: 142530336 - 142550151 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083312.2 NP_001076781.1 8 - 9 1475 60
NM_145545.4 NP_663520.2 8 - 9 1493 60
GenBank mRNA AK036666.1 - 8 - 9 1489 60
AK087751.1 - 8 - 9 1469 60
AK131176.1 - 7 - 8 1376 60
BC010229.1 - 8 - 9 1440 60

View Full Product Details