Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145585.2 | NP_663560.1 | 3 - 4 | 714 | 148 |
GenBank mRNA | AK032078.1 | - | 3 - 4 | 690 | 148 |
AK152912.1 | - | 3 - 4 | 687 | 148 | |
AK178112.1 | - | 1 - 2 | 140 | 148 | |
BC004776.1 | - | 3 - 4 | 681 | 148 |