Assay Design
Gene Transcripts
- Gene Symbol
- Serinc4
- Entrez Gene ID
-
574418
- Gene Name
- serine incorporator 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 121451177 - 121456764 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC026459.1 |
- |
6 - 7 |
965 |
64 |
View Full Product Details