Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190343.1 | NP_001177272.1 | 3 - 4 | 407 | 118 |
NM_001190344.1 | NP_001177273.1 | 4 - 5 | 493 | 118 | |
NM_146014.3 | NP_666126.1 | 4 - 5 | 581 | 118 | |
GenBank mRNA | BC029157.1 | - | 4 - 5 | 512 | 118 |