Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190343.1 | NP_001177272.1 | 6 - 7 | 744 | 68 |
NM_001190344.1 | NP_001177273.1 | 7 - 8 | 830 | 68 | |
NM_146014.3 | NP_666126.1 | 7 - 8 | 918 | 68 | |
GenBank mRNA | AK155145.1 | - | 7 - 8 | 892 | 68 |
AK166131.1 | - | 4 - 5 | 445 | 68 | |
AK169727.1 | - | 7 - 8 | 832 | 68 | |
AY442689.1 | - | 7 - 8 | 805 | 68 | |
BC029157.1 | - | 7 - 8 | 849 | 68 |