Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146105.3 | NP_666217.2 | 10 - 11 | 2072 | 65 |
GenBank mRNA | AK036332.1 | - | 10 - 11 | 2104 | 65 |
AK146681.1 | - | 10 - 11 | 2162 | 65 | |
BC034107.1 | - | 3 - 4 | 1090 | 65 | |
BC039206.1 | - | 8 - 9 | 1435 | 65 | |
BC054802.1 | - | 10 - 11 | 2072 | 65 |