Assay Design



Gene Transcripts

Gene Symbol
Mybpc2
Entrez Gene ID
233199
Gene Name
myosin binding protein C, fast-type
Gene Aliases
-
Chromosome Location
Chr.7: 44501699 - 44524715 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146189.3 NP_666301.2 3 - 4 264 59
GenBank mRNA AK029282.1 - 3 - 4 250 59
AK132563.1 - 3 - 4 252 59
AK161146.1 - 3 - 4 252 59
BC029762.1 - 3 - 4 234 59
BC039184.1 - 3 - 4 229 59
CK787764.1 - 3 - 4 278 59

View Full Product Details