Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146189.3 | NP_666301.2 | 20 - 21 | 2405 | 70 |
GenBank mRNA | AK029282.1 | - | 20 - 21 | 2391 | 70 |
AK132563.1 | - | 20 - 21 | 2395 | 70 | |
AK161146.1 | - | 20 - 21 | 2393 | 70 | |
BC029762.1 | - | 20 - 21 | 2375 | 70 | |
BC038869.1 | - | 5 - 6 | 668 | 70 | |
BC039184.1 | - | 20 - 21 | 2370 | 70 |