Assay Design



Gene Transcripts

Gene Symbol
Mybpc2
Entrez Gene ID
233199
Gene Name
myosin binding protein C, fast-type
Gene Aliases
-
Chromosome Location
Chr.7: 44501699 - 44524715 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146189.3 NP_666301.2 27 - 28 3478 78
GenBank mRNA AK029282.1 - 27 - 28 3464 78
AK132563.1 - 27 - 28 3469 78
AK161146.1 - 27 - 28 3468 78
BC029762.1 - 27 - 28 3448 78
BC038869.1 - 12 - 13 1741 78
BC039184.1 - 27 - 28 3443 78

View Full Product Details