Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021492.3 | NP_067467.2 | 1 - 2 | 412 | 83 |
GenBank mRNA | AK134504.1 | - | 1 - 2 | 161 | 83 |
AK138496.1 | - | 1 - 2 | 271 | 83 | |
AY528675.1 | - | 1 - 2 | 193 | 83 | |
BC024722.1 | - | 1 - 2 | 142 | 83 | |
BC139378.1 | - | 1 - 2 | 160 | 83 | |
BC139379.1 | - | 1 - 2 | 160 | 83 | |
CJ064603.1 | - | 1 - 2 | 413 | 83 |