Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146219.1 | NP_666331.1 | 4 - 5 | 1452 | 117 |
GenBank mRNA | AK131135.1 | - | 4 - 5 | 1377 | 117 |
AK157027.1 | - | 4 - 5 | 1486 | 117 | |
AK164831.1 | - | 4 - 5 | 1487 | 117 | |
BC025816.1 | - | 4 - 5 | 1452 | 117 |