Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163493.1 | NP_001156965.1 | 13 - 14 | 3379 | 76 |
NM_146258.2 | NP_666370.3 | 13 - 14 | 3322 | 76 | |
GenBank mRNA | AK050259.1 | - | 13 - 14 | 3380 | 76 |
AK136202.1 | - | 9 - 10 | 1905 | 76 | |
BC027830.1 | - | 13 - 14 | 3334 | 76 |