Assay Design
Gene Transcripts
- Gene Symbol
- Fxyd2
- Entrez Gene ID
-
11936
- Gene Name
- FXYD domain-containing ion transport regulator 2
- Gene Aliases
-
Atp1g1
- Chromosome Location
-
Chr.9: 45399709 - 45410278 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_028499.1 |
|
1 - 2 |
296 |
74 |
GenBank mRNA |
CA491597.1 |
- |
1 - 2 |
330 |
74 |
View Full Product Details