Product Details
- Assay Reference Genome
Location
Chr.1:119053409 on build GRCm38
- Cytoband
- 1E2.3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 14633
- Gene Symbol
- Gli2
- Gene Name
- GLI-Kruppel family member GLI2
- Gene Aliases
- AW546128
- Location
- Chr.1:118834061-119053619 on build GRCm38
- Assay Gene Location
- Within Exon 1
View Full Product Details