Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007426.4 | NP_031452.2 | 8 - 9 | 1612 | 74 |
GenBank mRNA | AF004326.1 | - | 8 - 9 | 1538 | 74 |
AK019860.1 | - | 8 - 9 | 1601 | 74 | |
AK048622.1 | - | 8 - 9 | 1569 | 74 | |
AK143974.1 | - | 8 - 9 | 1600 | 74 | |
AK156132.1 | - | 8 - 9 | 1583 | 74 | |
BC027216.1 | - | 8 - 9 | 1565 | 74 | |
BC138309.1 | - | 8 - 9 | 1506 | 74 | |
BC138313.1 | - | 8 - 9 | 1506 | 74 |