Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007440.4 | NP_031466.2 | 11 - 12 | 1606 | 73 |
GenBank mRNA | AK036898.1 | - | 11 - 12 | 1604 | 73 |
AK036960.1 | - | 11 - 12 | 1562 | 73 | |
AK087724.1 | - | 11 - 12 | 1607 | 73 | |
AK172216.1 | - | 11 - 12 | 1604 | 73 | |
AK172488.1 | - | 11 - 12 | 1607 | 73 | |
BC152329.1 | - | 11 - 12 | 1611 | 73 | |
BC152403.1 | - | 11 - 12 | 1611 | 73 | |
S80446.1 | - | 11 - 12 | 1618 | 73 | |
U04334.1 | - | 11 - 12 | 1537 | 73 |