Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007462.3 | NP_031488.2 | 9 - 10 | 1235 | 68 |
GenBank mRNA | AK045053.1 | - | 9 - 10 | 1206 | 68 |
AK046280.1 | - | 9 - 10 | 1225 | 68 | |
AK080907.1 | - | 9 - 10 | 1235 | 68 | |
BC141141.1 | - | 8 - 9 | 948 | 68 | |
M88127.1 | - | 8 - 9 | 924 | 68 |