Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007462.3 | NP_031488.2 | 15 - 16 | 2267 | 104 |
GenBank mRNA | AK046280.1 | - | 15 - 16 | 2257 | 104 |
AK080907.1 | - | 15 - 16 | 2267 | 104 | |
BC141141.1 | - | 14 - 15 | 1980 | 104 | |
M88127.1 | - | 14 - 15 | 1956 | 104 |