Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009963.4 | NP_034093.1 | 1 - 2 | 255 | 70 |
GenBank mRNA | AB003433.1 | - | 1 - 2 | 140 | 70 |
AF156987.1 | - | 1 - 2 | 223 | 70 | |
AK037778.1 | - | 1 - 2 | 223 | 70 | |
AK041696.1 | - | 1 - 2 | 223 | 70 | |
AK133781.1 | - | 1 - 2 | 230 | 70 | |
AK172994.1 | - | 1 - 2 | 187 | 70 | |
BC054794.1 | - | 1 - 2 | 211 | 70 | |
BC066799.1 | - | 1 - 2 | 214 | 70 | |
CJ164467.1 | - | 1 - 2 | 255 | 70 |