Assay Design
Gene Transcripts
- Gene Symbol
- Tsc22d2
- Entrez Gene ID
-
72033
- Gene Name
- TSC22 domain family, member 2
- Gene Aliases
-
1810043J12Rik, 5530402M19Rik
- Chromosome Location
-
Chr.3: 58415531 - 58466787 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK007760.1 |
- |
1 - 2 |
340 |
99 |
BC058221.1 |
- |
1 - 2 |
193 |
99 |
BC117998.1 |
- |
2 - 3 |
1726 |
99 |
View Full Product Details