Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039692.1 | NP_001034781.1 | 4 - 5 | 1348 | 81 |
NM_029277.2 | NP_083553.2 | 4 - 5 | 1348 | 81 | |
GenBank mRNA | AK012717.2 | - | 4 - 5 | 1190 | 81 |
AK031648.1 | - | 4 - 5 | 1192 | 81 | |
AK077063.1 | - | 4 - 5 | 1349 | 81 | |
BC037608.1 | - | 4 - 5 | 1163 | 81 | |
BC141184.1 | - | 4 - 5 | 1157 | 81 | |
BC145506.1 | - | 4 - 5 | 1156 | 81 |