Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172116.4 | NP_742114.3 | 2 - 3 | 204 | 77 |
GenBank mRNA | AK081187.1 | - | 2 - 3 | 164 | 77 |
AK084273.1 | - | 2 - 3 | 160 | 77 | |
AK088159.1 | - | 2 - 3 | 180 | 77 | |
AK140975.1 | - | 2 - 3 | 155 | 77 | |
AK161205.1 | - | 2 - 3 | 170 | 77 | |
BC023835.1 | - | 2 - 3 | 174 | 77 | |
BY118625.1 | - | 2 - 3 | 204 | 77 |