Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172142.3 | NP_742154.1 | 6 - 7 | 910 | 84 |
GenBank mRNA | AK155969.1 | - | 7 - 8 | 916 | 84 |
AK167356.1 | - | 7 - 8 | 904 | 84 | |
AY078069.1 | - | 6 - 7 | 915 | 84 | |
BC110633.1 | - | 6 - 7 | 870 | 84 |