Assay Design



Gene Transcripts

Gene Symbol
Zmynd8
Entrez Gene ID
228880
Gene Name
zinc finger, MYND-type containing 8
Gene Aliases
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7
Chromosome Location
Chr.2: 165784152 - 165898984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252584.2 NP_001239513.1 3 - 4 451 72
NM_001252587.2 NP_001239516.1 4 - 5 463 72
NM_001281926.1 NP_001268855.1 3 - 4 350 72
GenBank mRNA AK147620.1 - 3 - 4 351 72
AK155864.1 - 3 - 4 256 72
BC023300.1 - 3 - 4 318 72
BC063267.1 - 3 - 4 244 72
CN536818.1 - 3 - 4 486 72
CN670238.1 - 4 - 5 278 72

View Full Product Details