Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172424.4 | NP_766012.3 | 2 - 3 | 376 | 76 |
GenBank mRNA | AK088795.1 | - | 2 - 3 | 419 | 76 |
AK142391.1 | - | 2 - 3 | 391 | 76 | |
AK154895.1 | - | 2 - 3 | 356 | 76 | |
AY338464.1 | - | 2 - 3 | 353 | 76 |