Assay Design



Gene Transcripts

Gene Symbol
Tbx4
Entrez Gene ID
21387
Gene Name
T-box 4
Gene Aliases
3930401C23
Chromosome Location
Chr.11: 85886343 - 85916099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011536.2 NP_035666.2 2 - 3 296 73
GenBank mRNA AB221665.1 - 2 - 3 296 73
AY075134.1 - 2 - 3 296 73

View Full Product Details