Assay Design



Gene Transcripts

Gene Symbol
Clic3
Entrez Gene ID
69454
Gene Name
chloride intracellular channel 3
Gene Aliases
2300003G24Rik
Chromosome Location
Chr.2: 25456843 - 25458776 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027085.3 NP_081361.1 1 - 2 71 60
GenBank mRNA AK009020.1 - 1 - 2 73 60
BC060967.1 - 1 - 2 76 60

View Full Product Details